Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MELMDANLCQ VIQMELDHER MSYLLYQMLC GIKHLHSAGI IHRDLKPSNI VVKSDCTLKI LDFGLARTAG TSFMMTPYVV TRYYRAPEVI LGMGYKENVD IWSVGCIMGE MVRHKILFPG RDYIDQWNKV IEQLGTPCPE FMKKLQPTVR NYVENRPKYA GLTFPKLFPD SLFPADSEHN KLKASQARDL LSKMLVIDPA KRISVDDALQ HPYINVWYDP AEVEAPPPQI YDKQLDEREH TIEEWKELIY KEVMNSEEKT KNGVVKGQPS PSGAAVNSSE SLPPSSSVND ISSMSTDQTL ASDTDSSLEA SAGPLGCCR
MAPK10 (JNK3) is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). MAPK10 is involved in neuronal apoptosis and phosphorylates MAP2K4. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This protein is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). Through its phosphorylation and nuclear localization, this kinase plays regulatory roles in the signaling pathways during neuronal apoptosis. Beta-arrestin 2, a receptor-regulated MAP kinase scaffold protein, is found to interact with, and stimulate the phosphorylation of this kinase by MAP kinase kinase 4 (NhpK4). Cyclin-dependent kianse 5 can phosphorylate, and inhibit the activity of this kinase, which may be important in preventing neuronal apoptosis. Four alternatively spliced transcript variants encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Jun N-terminal kinase 3; JNK3 alpha protein kinase; MAP kinase 10; MAP kinase p49 3F12; MAPK 10; Mitogen-activated protein kinase 10; OTTHUMP00000161181; OTTHUMP00000161183; OTTHUMP00000218718; SAPK1b; stress activated protein kinase beta; Stress-activated protein kinase 1b; Stress-activated protein kinase JNK3
Gene Aliases: JNK3; JNK3A; MAPK10; p493F12; p54bSAPK; PRKM10; SAPK1B
UniProt ID: (Human) P53779
Entrez Gene ID: (Human) 5602
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support