Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MAPK13 (p38 delta) is a serine/threonine kinase that transduces a variety of extracellular signals, including cytokine and environmental stress responses. MAPK13 also regulates the differentiation of keratinocytes. This kinase is activated by various environmental stresses and proinflammatory cytokines. The activation requires its phosphorylation by MAP kinase kinases, or its autophosphorylation triggered by the interaction of MAP3K7IP1/TAB1 protein with this kinase. The substrates of this kinase include transcription regulator ATF2, MEF2C, and MAX, cell cycle regulator CDC25B, and tumor suppressor p53, which suggest the roles of this kinase in stress related transcription and cell cycle regulation, as well as in genotoxic stress response. Four alternatively spliced transcript variants of SAPK4 encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAP kinase 13; MAP kinase p38 delta; MAPK 13; mitogen activated protein kinase 13; Mitogen-activated protein kinase 13; Mitogen-activated protein kinase p38 delta; p38 delta MAP kinase; SAPK/Erk/kinase 4; Stress-activated protein kinase 4
Gene Aliases: MAPK 13; MAPK-13; MAPK13; p38delta; PRKM13; SAPK4; Serk4
UniProt ID: (Human) O15264, (Mouse) Q9Z1B7
Entrez Gene ID: (Dog) 612821, (Human) 5603, (Rat) 29513, (Mouse) 26415
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support