Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Predicted reactivity by homology: Orangutan, Bovine.
Mitogen-activated protein kinase 1-interacting protein 1-like (MAPK1IP1L) belongs to the MISS family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAPK-interacting and spindle-stabilizing protein; MAPK-interacting and spindle-stabilizing protein-like; mitogen activated protein kinase 1 interacting protein 1-like; mitogen-activated protein kinase 1 interacting protein 1-like; Mitogen-activated protein kinase 1-interacting protein 1-like
Gene Aliases: 9130202B12; C130032J12Rik; c14_5346; C14orf32; MAPK1IP1L; MISS
UniProt ID: (Human) Q8NDC0, (Mouse) Q8BH93
Entrez Gene ID: (Human) 93487, (Mouse) 218975
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support