Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The antibody detects a 32 kDa* prtoein corresponding to the molecular mass of EB3 on SDS-PAGE immunoblots of human HeLa cells and mouse brain tissue. The antibody also detects EB3 by immunocytochemistry in HeLa and A431 cells.
EB3 is a 282 amino acid homologue of EB1, a protein known to bind APC (adenomatous polyposis coli). It has been shown to bind to tubulin in vitro and microtubules in vivo. Northern blotting shows expressions to be mainly in brain, and to a lesser degree in muscle. EB3 interacts with APCL, a homologue of the tumor suppressor APC, and this interaction may be involved in the stabilization or extension of microtubules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APC binding protein; EB1 protein family member 3; EB3; EBF3; End-binding protein 3; Microtubule-associated protein RP/EB family member 3; microtubule-associated protein, RP/EB family, member 3; RP3
Gene Aliases: AI790651; EB2; EB3; EBF3; EBF3-S; MAPRE3; RP3
UniProt ID: (Human) Q9UPY8, (Mouse) Q6PER3, (Rat) Q5XIT1
Entrez Gene ID: (Human) 22924, (Mouse) 100732, (Rat) 298848
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support