Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPDQALQQM LDRSCWVCFA TDEDDRTAEW VRPCRCRGST KWVHQACLQR WVDEKQRGNS TARVACPQCN AEYLIVFPKL GPVVYVLDLA DRLISKACPF AAAGIMVGSI YWTAVTYGAV TVMQVVGHKE GLDVMERADP LFLLIGLPTI PVMLILGKMI RWEDYVLRLW RKYSNKLQIL NSIFPGIGCP VPRIPAEANP LADHVSATRI (1-209 aa encoded by BC015480)
MARCH5 is a ubiquitin ligase of the mitochondrial outer membrane that plays a role in the control of mitochondrial morphology by regulating mitofusin-2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase MARCHF5; MARCH-V; membrane associated ring finger 5; membrane-associated ring finger (C3HC4) 5; Membrane-associated RING finger protein 5; Membrane-associated RING-CH protein V; Mitochondrial ubiquitin ligase; MITOL; RING finger protein 153; RING-type E3 ubiquitin transferase MARCHF5
Gene Aliases: MARCH-V; MARCH5; MARCHF5; MITOL; RNF153
UniProt ID: (Human) Q9NX47
Entrez Gene ID: (Human) 54708
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support