Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: WYSESEITQG ARSRSQNQQR DHDSKRPKLS CTNCTTSAGR NVGNGLNTLS DSSWRHSQVP RSSSMVLGSF G
The function of protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Axotrophin; C3HC4 7; DKFZP586F1122; E3 ubiquitin-protein ligase MARCH7; E3 ubiquitin-protein ligase MARCHF7; gene trap ROSA b-geo 17; MARCH-VII; membrane associated ring finger 7; membrane-associated ring finger (C3HC4) 7, E3 ubiquitin protein ligase; Membrane-associated RING finger protein 7; Membrane-associated RING-CH protein VII; RING finger protein 177; RING-type E3 ubiquitin transferase MARCHF7
Gene Aliases: AXO; AXOT; Gtrgeo17; MARCH-VII; MARCH7; MARCHF7; RNF177
UniProt ID: (Human) Q9H992, (Rat) Q5XI50, (Mouse) Q9WV66
Entrez Gene ID: (Human) 64844, (Rat) 311059, (Mouse) 57438
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support