Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: HHKVQRSVFS SQKQRRYSDH AGPAIPSVVA YPKRSQTSTA DSDLKEDGIS SRKSSGSAVG GKGIAPASPM LGNASNPNKA DIPERKKSST VPSSNTASG
MARK3 is a member of a group of kinases known to regulate cell polarity via phosphorylation of microtubule-associated proteins (MAPs). The protein encoded by MARK3 is activated by phosphorylation and in turn is involved in the phosphorylation of tau proteins MAP2 and MAP4. Several transcript variants encoding different isoforms have been found for MARK3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-TAK1; Cdc25C-associated protein kinase 1; cTAK1; ELKL motif kinase 2; EMK-2; MAP/microtubule affinity-regulating kinase 3; Par-1a; Protein kinase STK10; Ser/Thr protein kinase PAR-1; Serine/threonine-protein kinase p78
Gene Aliases: CTAK1; EMK2; KP78; MARK3; Par-1a; PAR1A
UniProt ID: (Human) P27448
Entrez Gene ID: (Human) 4140
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support