Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Immunohistochemistry (PFA fixed): incubate antibody for 30 min RT. Staining of formalin-fixed tissues requires heating tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 45 min at 95 degrees C followed by cooling at RT for 20 minutes.
MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antigen LB39-AA; Antigen SK29-AA; MART-1; melanoma antigen recognized by T-cells; Melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; Protein Melan-A; tumor associated antigen
Gene Aliases: A930034P04Rik; MART-1; MART1; MLANA
UniProt ID: (Human) Q16655
Entrez Gene ID: (Human) 2315, (Dog) 403495, (Mouse) 77836, (Rat) 293890
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support