Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product is supplied at 1 mL (100 µL/mL).
Application Notes: This product can be used as a detector in immunoassays.
MBL is a key component in immune response in that it can directly trigger neutralization of invading microorganisms by an Ab-independent mechanism. Mutations of human MBL are associated with immunodeficiency resulting from a reduction in the ability of the mutant MBL to initiate complement fixation. In human, three types of MBL-associated serine proteases, MASP-1, MASP-2 and MASP-3, and a truncated form of MASP-2 (small MBL-associated protein; sMAP or MAp19) complex with MBL to activate the lectin pathway of the complement system. MASP-3 is an alternatively spliced product from the MASP-1 gene. The heavy/A chains are identical between MASP-1 and MASP-3 but the light/B chains are entirely different. Activated MASPs subsequently cleave and activate downstream components of the complement pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C4/C2 activating component of Ra-reactive factor; Complement factor MASP-3; Complement-activating component of Ra-reactive factor; mannan-binding; mannan-binding lectin serine peptidase 1 (C4/C2 activating component of Ra-reactive factor); Mannan-binding lectin serine protease 1; Mannose-binding lectin-associated serine protease 1; Mannose-binding protein-associated serine protease; MASP-1; Ra-reactive factor serine protease p100; RaRF; Serine protease 5
Gene Aliases: 3MC1; CRARF; CRARF1; MAP1; MAp44; MASP; MASP1; MASP3; PRSS5; RaRF
UniProt ID: (Human) P48740
Entrez Gene ID: (Human) 5648
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support