Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is mouse liver tissue lysate.
PA5-20725 can be used with blocking peptide PEP-0839.
This gene catalyzes a two-step reaction that involves the transfer of the adenosyl moiety of ATP to methionine to form S-adenosylmethionine and tripolyphosphate, which is subsequently cleaved to PPi and Pi. S-adenosylmethionine is the source of methyl groups for most biological methylations. The encoded protein is found as a homotetramer or a homodimer whereas a third form, MAT II , is encoded by the MAT2A gene. Mutations in this gene are associated with methionine adenosyltransferase deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cartilage matrix protein; MAT1A; MATA1; matrilin 1, cartilage matrix protein 1; Matrilin-1; SAMS; SAMS1
Gene Aliases: CMP; CRTM; Mat1; MATN1; matrilin-1
UniProt ID: (Human) P21941, (Mouse) P51942
Entrez Gene ID: (Human) 4146, (Rat) 297894, (Mouse) 17180
Molecular Function: extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support