Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GRVIHLSNL PHSGYSDSAV LKLAEPYGKI KNYILMRMKS QAFIEMETRE DAMAMVDHCL KKALWFQGRC VKVDLSEKYK KLVLRIPNRG IDLLKKDKSR KRSYSPDGKE SPSDKKSKTD GSQKTESSTE GKEQEEKSGE DGEKDTKDDQ TEQEPNMLLE SEDELLVDEE EAAALLESGS SVGDETDLAN LGDVASDGKK EPSDKAVKKD GSASAAAKKK LKKVDKIEEL DQENEAALEN GIKNEENTEP GAESSENADD PNKDTSENAD GQSDENKDDY TIPDEYRIGP YQPNVPVGID YVIPKTGFYC KLCSLFYTNE EVAKNTHCSS LPHYQKLKKF LNKLAEERRQ KKET (495-848 aa encoded by B C015031)
This gene encodes a nuclear matrix protein, which is proposed to stabilize certain messenger RNA species. Mutations of this gene are associated with distal myopathy 2, which often includes vocal cord and pharyngeal weakness. Alternatively spliced transcript variants, including read-through transcripts composed of the upstream small nucleolar RNA host gene 4 (non-protein coding) and matrin 3 gene sequence, have been identified. Pseudogenes of this gene are located on chromosomes 1 and X.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Matrin-3; Nuclear scaffold protein p130/MAT3; vocal cord and pharyngeal weakness with distal myopathy
Gene Aliases: 1110061A14Rik; 2810017I02Rik; AI841759; ALS21; AW555618; D030046F20Rik; KIAA0723; MATR3; mKIAA0723; MPD2; P130/MAT3; VCPDM
UniProt ID: (Human) P43243, (Mouse) Q8K310, (Rat) P43244
Entrez Gene ID: (Human) 9782, (Mouse) 17184, (Rat) 29150
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support