Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPLNQILNQ HNAASFPASS LLSAAAKAQL ANQNKLAGNN SSSSSNSGAV AGSGNTEGHS TLNTMFPPTA NMLLPTGEGQ SGRAALRDKL MSQQKDALRK RKQPPTTVLS LLRQSQMDSS AVPKPGPDLL RKQGQGSFPI SSMSQLLQSM SCQSSHLSSN STPGCGASNT ALPCSANQLH FTDPSMNSSV LQNIPLRGEA VHCHNANTNF VHSNSPVPNH HLAGLINQIQ ASGNCGMLSQ SGMALGNSLH PNPPQSRIST SSTPVIPNSI VSSYNQTSSE AGMVLLEKST QRY (1-292 aa encoded by BC014534)
MBD5 binds to heterochromatin but does not interact with either methylated or unmethylated DNA (in vitro).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ11113; FLJ30517; Methyl-CpG-binding domain protein 5; Methyl-CpG-binding protein MBD5
Gene Aliases: 9430004D19Rik; AA536666; AI426407; C030040A15Rik; Gm1630; KIAA1461; MBD5; MRD1; OTTMUSG00000012483
UniProt ID: (Human) Q9P267, (Mouse) B1AYB6
Entrez Gene ID: (Human) 55777, (Mouse) 109241, (Rat) 311026
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support