Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-type lectin; Collectin1; mannan-binding lectin; Mannan-binding protein; mannose binding lectin 2 (protein C); mannose binding lectin, liver (A); mannose-binding lectin (protein C) 2, soluble; Mannose-binding protein; Mannose-binding protein C; mannose-binding protein C (liver); MBP; MBP-C; MBPC; MGC116832; MGC116833; RA-reactive factor P28A subunit; Ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A
Gene Aliases: Ab2-001; Ab2-011; cMBl; collectin; MBL; Mbl2; MBP
UniProt ID: (Chicken) Q98TA4, (Rat) P08661
Entrez Gene ID: (Chicken) 374267, (Rat) 64668
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support