Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody stains mouse Melanocortin 3R/MC3R-transfected cells but not irrelevant transfectants.
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a G-protein-coupled receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone that is expressed in tissues other than the adrenal cortex and melanocytes. This gene maps to the same region as the locus for benign neonatal epilepsy. Mice deficient for this gene have increased fat mass despite decreased food intake, suggesting a role for this gene product in the regulation of energy homeostasis. Mutations in this gene are associated with a susceptibility to obesity in humans.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MC3 receptor; MC3-R; Melanocortin receptor 3
Gene Aliases: MC3-R; Mc3r
UniProt ID: (Mouse) P33033
Entrez Gene ID: (Mouse) 17201
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support