Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ALASLKGDIV ELNKRLQQTE RERDLLEKKL AKAQCEQSHL MREHEDVQER TTLRYEERIT ELHSVIAELN KKIDRLQGTT IREEDEYSEL RSEL
Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
This gene is a candidate colorectal tumor suppressor gene that is thought to negatively regulate cell cycle progression. The orthologous gene in the mouse expresses a phosphoprotein associated with the plasma membrane and membrane organelles, and overexpression of the mouse protein inhibits entry into S phase. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Colorectal mutant cancer protein; Protein MCC
Gene Aliases: D18Ertd451e; E330037C19; MCC; MCC1
UniProt ID: (Human) P23508
Entrez Gene ID: (Human) 4163, (Mouse) 328949
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support