Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQDIAFLSGG RGKDNAWIIT FPENCNFRCI PEEVIAKVLT YLTSIARQNG SDSRFTIILD RRLDTWSSLK ISLQKISASF PGNLHLVLVL RPTSFLQRTF TDIGFWFSQE DFMLKLPVVM LSSVSDLLTY IDDKQLTPEL GGTLQYCHSE WIIFRNAIEN FALTVKEMAQ MLQSFGTELA ETELPDDIPS IEEILAIRAE RYHLLKNDIT AVTKEGKILL TNLEVPDTEG AVSSRLECHR QISGDWQTIN KLLTQVHDME TAFDGFWEKH QLKMEQYLQL WKFEQDFQQL VTEVEFLLNQ QAELADVTGT IAQVKQKIKK LENLDENSQE LLSKAQFVIL HGHKLAANHH YALDLICQRC NELRYLSDIL VNEIKAKRIQ LSRTFKMHKL LQQARQCCDE GECLLANQEI DKFQSKEDAQ KALQDIENFL EMALPFINYE PETLQYEFDV ILSPELKVQM KTIQLKLENI RSIFENQQAG FRNLADKHVR PIQFVVPTPE NLVTSGTPFF SSKQGKKTWR QNQSNLKIEV VPDCQEKRSS GPSSSLDNGN SLDVLKNHVL NELIQTERVY VRELYTVLLG YRAEMDNPEM FDLMPPLLRN KKDILFGNMA EIYEFHNDIF LSSLENCAHA PERVGPCFLE RKDDFQMYAK YCQNKPRSET IWRKYSECAF FQECQRKLKH RLRLDSYLLK PVQRITKYQL LLKELLKYSK DCEGSALLKK ALDAMLDLLK SVNDSMHQIA INGYIGNLNE LGKMIMQGGF SVWIGHKKGA TKMKDLARFK PMQRHLFLYE KAIVFCKRRV ESGEGSDRYP SYSFKHCWKM DEVGITEYVK GDNRKFEIWY GEKEEVYIVQ ASNVDVKMTW LKEIRNILLK QQELLTVKKR KQQDQLTERD KFQISLQQND EKQQGAFIST EETELEHTST VVEVCEAIAS VQAEANTVWT EASQSAEISE EPAEWSSNYF YPTYDENEEE NRPLMRPVSE MALLY
Guanine nucleotide exchange factor (GEF) that modulates the Rho family of GTPases. Promotes the conversion of some member of the Rho family GTPase from the GDP-bound to the GTP-bound form. Isoform 1 exhibits no activity toward RHOA, RAC1 or CDC42. Isoform 2 exhibits decreased GEF activity toward CDC42. Isoform 3 exhibits a weak but significant activity toward RAC1 and CDC42. Isoform 4 exhibits significant activity toward RHOA and CDC42. The truncated DBL oncogene is active toward RHOA, RAC1 and CDC42.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Oncogene MCF2 (oncogene DBL); OTTHUMP00000024159; Proto-oncogene DBL; Proto-oncogene MCF-2
Gene Aliases: ARHGEF21; DBL; MCF2
UniProt ID: (Human) P10911
Entrez Gene ID: (Human) 4168
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support