Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FDILCVVRD TVDPVQDEML ARFVVGSHVR HHPSNKEEEG LANGSAAEPA MPNTYGVEPL PQEVLKKYII YAKERVHPKL NQMDQDKVAK MYSDLRKESM ATGSIPITVR HIESMIRMAE AHARIHLRDY VIEDDVNMAI RVMLESFIDT QKFSVMRSMR KTFARYLSFR RDNNELLLFI LKQLVAEQVT YQRNRFGAQQ DTIEVPEKDL VDKARQINIH NLSAFYDSEL FRMNKFSHDL KRKMILQQF (657-904 aa encoded by BC007670)
MCM2 (Mini chromosome maintenance protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell devision cycle-like 1; cyclin-like 1; DNA replication licensing factor MCM2; MGC10606; mini chromosome maintenance deficient 2; minichromosome maintenance deficient 2 (mitotin); minichromosome maintenance deficient 2 mitotin; Minichromosome maintenance protein 2 homolog; Nuclear protein BM28
Gene Aliases: AA959861; AW476101; BM28; CCNL1; cdc19; CDCL1; D3S3194; DFNA70; KIAA0030; MCM2; Mcmd2; MITOTIN; mKIAA0030
UniProt ID: (Human) P49736, (Mouse) P97310
Entrez Gene ID: (Human) 4171, (Rat) 312538, (Mouse) 17216
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support