Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IQEMPEKAP AGQLPRSVDV ILDDDLVDKA KPGDRVQVVG TYRCLPGKKG GYTSGTFRTV LIACNVKQMS KDAQPSFSAE DIAKIKKFSK TRSKDIFDQL AKSLAPSIHG HDYVKKAILC LLLGGVERDL ENGSHIRGDI NILLIGDPSV AKSQLLRYVL CTAPRAIPTT GRGSSGVGLT AAVTTDQETG ERRLEAGAMV LADRGVVCID EFDKMSDMDR TAIHEVMEQG RVTIAKAGIH ARLNARCSVL AAANPVYGRY DQYKTPMENI GLQDSLLSRF DLLFIMLDQM DPEQDREISD HVLR (201-503 aa encoded by B C001626)
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein is a subunit of the protein complex that consists of MCM2-7. It has been shown to interact directly with MCM5/CDC46. This protein also interacts with, and thus is acetlyated by MCM3AP, a chromatin-associated acetyltransferase. The acetylation of this protein inhibits the initiation of DNA replication and cell cycle progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cervical cancer proto-oncogene 5; DNA polymerase alpha holoenzyme-associated protein P1; DNA replication factor MCM3; DNA replication licensing factor MCM3; hRlf beta subunit; MCM3 minichromosome maintenance deficient 3; MGC1157; mini chromosome maintenance deficient; minichromosome maintenance deficient 3; P1-MCM3; p102; replication licensing factor, beta subunit; RLF subunit beta; RP1-108C2.3
Gene Aliases: AL033361; C80350; HCC5; MCM3; Mcmd; Mcmd3; P1; P1-MCM3; P1.h; p1.m; RLFB
UniProt ID: (Human) P25205, (Mouse) P25206
Entrez Gene ID: (Human) 4172, (Mouse) 17215, (Rat) 316273
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support