Product References
Tumor Cell-Autonomous Pro-Metastatic Activities of PD-L1 in Human Breast Cancer Are Mediated by PD-L1-S283 and Chemokine Axes.
Cancers
Erlichman N,Baram T,Meshel T,Morein D,Da'adoosh B,Ben-Baruch A
500-P34Bt was used in Enzyme-linked immunosorbent assay to investigate the role of WT-PD-L1 in triple-negative breast cancer.
Fri Feb 18 00:00:00 EST 2022
Tumor-Stroma-Inflammation Networks Promote Pro-metastatic Chemokines and Aggressiveness Characteristics in Triple-Negative Breast Cancer.
Frontiers in immunology
Liubomirski Y,Lerrer S,Meshel T,Rubinstein-Achiasaf L,Morein D,Wiemann S,Körner C,Ben-Baruch A
500-P34Bt was used in Enzyme-linked immunosorbent assay to study the impact of tumor-stroma-inflammation networks on pro-metastatic phenotypes in triple-negative breast cancer (TNBC) to identify a novel tumor-stroma-inflammation networks that may promote TNBC aggressiveness by increasing the pro-malignancy potential of the TME and of the tumor cells themselves, and reveal key roles for CXCL8 in mediating these metastasis-promoting activities.
Mon Aug 10 00:00:00 EDT 2020
Co-Inflammatory Roles of TGFβ1 in the Presence of TNFα Drive a Pro-inflammatory Fate in Mesenchymal Stem Cells.
Frontiers in immunology
Lerrer S,Liubomirski Y,Bott A,Abnaof K,Oren N,Yousaf A,Körner C,Meshel T,Wiemann S,Ben-Baruch A
500-P34Bt was used in Enzyme-linked immunosorbent assay to demonstrate a TNFα + TGFβ1-driven pro-inflammatory fate in MSCs, identify specific molecular mechanisms involved, and propose that TNFα + TGFβ1-stimulated MSCs influence the tumor niche.
Wed Nov 20 00:00:00 EST 2019
M-sec regulates polarized secretion of inflammatory endothelial chemokines and facilitates CCL2-mediated lymphocyte transendothelial migration.
Journal of leukocyte biology
Barzilai S,Blecher-Gonen R,Barnett-Itzhaki Z,Zauberman A,Lebel-Haziv Y,Amit I,Alon R
500-P34Bt was used in Enzyme-linked immunosorbent assay to the first evidence of a novel inflammation-inducible machinery that regulates polarized secretion of endothelial CCL2 and other inflammatory chemokines and cytokines into basolateral endothelial compartments and facilitates the ability of endothelial CCL2 to promote T cell transendothelial migration.
Wed Jun 01 00:00:00 EDT 2016
Regulation of the inflammatory profile of stromal cells in human breast cancer: prominent roles for TNF-α and the NF-κB pathway.
Stem cell research & therapy
Katanov C,Lerrer S,Liubomirski Y,Leider-Trejo L,Meshel T,Bar J,Feniger-Barish R,Kamer I,Soria-Artzi G,Kahani H,Banerjee D,Ben-Baruch A
500-P34Bt was used in Enzyme-linked immunosorbent assay to study the impact of tumour necrosis factor α and interleukin 1β on the inflammatory phenotype of cancer-associated fibroblasts and mesenchymal stem cells.
Fri May 01 00:00:00 EDT 2015
Breast cancer: coordinated regulation of CCL2 secretion by intracellular glycosaminoglycans and chemokine motifs.
Neoplasia (New York, N.Y.)
Lebel-Haziv Y,Meshel T,Soria G,Yeheskel A,Mamon E,Ben-Baruch A
500-P34Bt was used in Enzyme-linked immunosorbent assay to propose that targeting these chemokine regions may lead to reduced secretion of CCL2 by breast cancer cells (and potentially also by other malignant cells).
Mon Sep 01 00:00:00 EDT 2014
The inflammatory cytokine TNFα cooperates with Ras in elevating metastasis and turns WT-Ras to a tumor-promoting entity in MCF-7 cells.
BMC cancer
Leibovich-Rivkin T,Liubomirski Y,Meshel T,Abashidze A,Brisker D,Solomon H,Rotter V,Weil M,Ben-Baruch A
500-P34Bt was used in Enzyme-linked immunosorbent assay to determine the relative contribution of two processes to breast cancer progression: (1) Intrinsic events, such as activation of the Ras pathway and down-regulation of p53; (2) The inflammatory cytokines TNFα and IL-1β, shown in our published studies to be highly expressed in tumors of >80% of breast cancer patients with recurrent disease.
Thu Mar 06 00:00:00 EST 2014
Progression of luminal breast tumors is promoted by ménage à trois between the inflammatory cytokine TNFα and the hormonal and growth-supporting arms of the tumor microenvironment.
Mediators of inflammation
Weitzenfeld P,Meron N,Leibovich-Rivkin T,Meshel T,Ben-Baruch A
500-P34Bt was used in Enzyme-linked immunosorbent assay to indicate that TNF α can turn into a strong prometastatic factor, suggesting a paradigm shift in which clinically approved inhibitors of TNFα would be applied in breast cancer therapy.
Thu Jun 05 00:00:00 EDT 2014
Inflammatory factors of the tumor microenvironment induce plasticity in nontransformed breast epithelial cells: EMT, invasion, and collapse of normally organized breast textures.
Neoplasia (New York, N.Y.)
Leibovich-Rivkin T,Liubomirski Y,Bernstein B,Meshel T,Ben-Baruch A
500-P34Bt was used in Enzyme-linked immunosorbent assay to reveal novel pathways through which the inflammatory microenvironment may contribute to relapsed disease in breast cancer.
Sun Dec 01 00:00:00 EST 2013
Tumor-promoting circuits that regulate a cancer-related chemokine cluster: dominance of inflammatory mediators over oncogenic alterations.
Cancers
Leibovich-Rivkin T,Buganim Y,Solomon H,Meshel T,Rotter V,Ben-Baruch A
500-P34Bt was used in Enzyme-linked immunosorbent assay to use an unbiased model system to propose that inflammatory mediators of the tumor milieu have dominating roles over oncogenic modifications in dictating the expression of a pro-malignancy chemokine readout.
Fri Jan 20 00:00:00 EST 2012
Trans- but not cis-resveratrol impairs angiotensin-II-mediated vascular inflammation through inhibition of NF-κB activation and peroxisome proliferator-activated receptor-gamma upregulation.
Journal of immunology (Baltimore, Md. : 1950)
Rius C,Abu-Taha M,Hermenegildo C,Piqueras L,Cerda-Nicolas JM,Issekutz AC,Estañ L,Cortijo J,Morcillo EJ,Orallo F,Sanz MJ
Wed Sep 15 00:00:00 EDT 2010
STAT4 and STAT6 regulate systemic inflammation and protect against lethal endotoxemia.
The Journal of clinical investigation
Lentsch AB,Kato A,Davis B,Wang W,Chao C,Edwards MJ
Thu Nov 01 00:00:00 EST 2001