Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
AA Sequence of recombinant protein: QPDALNVPST CCFTFSSKKI SLQRLKSYVI TTSRCPQKAV IFRTKLGKEI CADPKEKWVQ NYMKHLGRKA HTLKT.
Preparation: Produced in mice immunized with highly pure Recombinant Human MCP-4 (CCL13). Anti-Human MCP-4 (CCL13)-specific antibody was purified from cell culture by Protein G affinity chromatography.
Sandwich ELISA: Assuming 100 µL/well, a concentration of 2.0-4.0 µg/mL of this antibody will detect at least 100 pg/mL of Recombinant Human MCP-4 (CCL13) when used with PeproTech Biotinylated Antigen Affinity Purified Anti-Human MCP-4 (CCL13) (500-P04GBT) as the detection antibody at a concentration of approximately 0.5-1.0 µg/mL.
Western Blot: To detect Human MCP-4 (CCL13) by Western Blot analysis this antibody can be used at a concentration of 0.25-0.50 µg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human MCP-4 (CCL13) is 0.5-1.0 ng/lane, under reducing or non-reducing conditions.
Monocyte Chemotactic Protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as IL-1 and TNFalpha. MCP-4 is a ligand for three different G protein coupled receptors, CCR2, CCR3 and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils and basophils and this signaling is associated with the allergic response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 13; CCL 13; chemokine (C-C motif) ligand 13; CK-beta-10; H-MCP-4; MCP-4; Monocyte chemoattractant protein 4; Monocyte chemotactic protein 4; NCC-1; new CC chemokine 1; small inducible cytokine subfamily A (Cys-Cys), member 13; Small-inducible cytokine A13
Gene Aliases: CCL13; CKb10; MCP-4; MCP4; NCC-1; NCC1; SCYA13; SCYL1
UniProt ID: (Human) Q99616
Entrez Gene ID: (Human) 6357
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support