Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: LO2, MCF7, Mouse heart, Mouse brain, Mouse liver, Mouse kidney; Cellular Location: Cytoplasm
Immunogen sequence: MEPEAPRRRH THQRGYLLTR NPHLNKDLAF TLEERQQLNI HGLLPPSFNS QEIQVLRVVK NFEHLNSDFD RYLLLMDLQD RNEKLFYRVL TSDIEKFMPI VYTPTVGLAC QQYSLVFRKP RGLFITIHDR GHIASVLNAW PEDVIKAIVV TDGERILGLG DLGCNGMGIP VGKLALYTAC GGMNPQECLP VILDVGTENE ELLKDPLYIG LRQRRVRGSE YDDFLDEFME AVSSKYGMNC LIQFEDFANV NAFRLLNKYR NQYCTFNDDI QGTASVAVAG LLAALRITKN KLSDQTILFQ
ME1 encodes a cytosolic, NADP-dependent enzyme that generates NADPH for fatty acid biosynthesis. The activity of this enzyme, the reversible oxidative decarboxylation of malate, links the glycolytic and citric acid cycles. The regulation of expression for this gene is complex. Increased expression can result from elevated levels of thyroid hormones or by higher proportions of carbohydrates in the diet.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytosolic malic enzyme 1; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); malate dehydrogenase (oxaloacetate-decarboxylating) (NADP+); Malic enzyme 1; malic enzyme 1, NADP(+)-dependent, cytosolic; malic enzyme 1, soluble; Malic enzyme, cytoplasmic; malic enzyme, supernatant; NADP-dependent malic enzyme; NADP-ME; pyruvic-malic carboxylase
Gene Aliases: D9Ertd267e; HUMNDME; Mdh-1; ME1; MES; Mod-1; MOD1
UniProt ID: (Human) P48163, (Mouse) P06801
Entrez Gene ID: (Human) 4199, (Mouse) 17436, (Rat) 24552
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support