Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive test controls include: HT-29, 293T, Mouse brain, Mouse heart, Mouse kidney, Rat brain, Rat heart, Rat kidney. The target is usually found in the following locations: Mitochondrion matrix.
Immunogen sequence: IRHIPDEIFL LTAEQIAQEV SEQHLSQGRL YPPLSTIRDV SLRIAIKVLD YAYKHNLASY YPEPKDKEAF VRSLVYTPDY DSFTLDSYTW PKEAMNVQTV
ME3 proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis, metastasis, and atherosclerosis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HME; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); Malic enzyme 3; malic enzyme 3, NADP(+)-dependent, mitochondrial; malic enzyme, NADP+-dependent, mitochondrial; mitochondrial malic enzyme 3; mitochondrial NADP(+)-dependent malic enzyme 3; MMP3; NADP-dependent malic enzyme, mitochondrial; NADP-ME; pyruvic-malic carboxylase
Gene Aliases: 1700020C08Rik; B230207H15Rik; ME3; NADP-ME
UniProt ID: (Human) Q16798, (Mouse) Q8BMF3
Entrez Gene ID: (Human) 10873, (Mouse) 109264, (Rat) 361602
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support