Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GQLIEPLQI FPRACKPPGE RNIHGLKVNT RAGPSQHSSP AVSDSLPSNS LKKSSAELKK ILANGQMNEQ DIRYRDTLGH GNGGTVYKAY HVPSGKILAV KVILLDITLE LQKQIMSELE ILYKCDSSYI IGFYGAFFVE NRISICTEFM DGGSLDVYRK MPEHVLGRIA VAVVKGLTYL WSLKILHRDV KPSNMLVNTR GQVKLCDFGV STQLVNSIAK TYVGTNAYMA PERISGEQYG IHSDVWSLGI SFMELALGRF PYPQIQKNQG SLMPLQLLQC IVDEDSPVLP VGEFSEPFVH FITQCMRKQP KERPAPEELM GHPFIVQFND GNAAVVSMWV CRALEERRSQ QGPP (96-448 aa encoded by BC008838)
MAP2K5 (MEK5) serine/threonine kinase (mitogen-activated protein kinase 5) or MEK5 is a dual specificity protein kinase that belongs to the MAP kinase family which specifically interacts with and activates MAPK7/ERK5. MAP2K5 itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). The signal cascade mediated by MAP2K5 is involved in growth factor stimulated cell proliferation and muscle cell differentiation. The rs3743354 polymorphism in the MAP2K5 promoter may affect the risk of developing colorectal cancer (CRC).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dual specificity mitogen-activated protein kinase kinase 5; MAP kinase kinase 5; MAP kinase kinase MEK5b; MAPK/ERK kinase 5; MAPK/ERK kinese 5; MAPKK 5; MEK 5; mitogen activated protein kinase kinase 5
Gene Aliases: AI324775; AI428457; HsT17454; MAP2K5; MAPKK5; MEK5; MKK5; PRKMK5
UniProt ID: (Human) Q13163, (Mouse) Q9WVS7, (Rat) Q62862
Entrez Gene ID: (Human) 5607, (Mouse) 23938, (Rat) 29568
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support