Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EDLISLWQY DHLTATYLLL LAKKARGKPV RLRLSSFSCG QASATPFTDI KSNNWSLEDV TASDKNYVAG LIDYDWCEDD LSTGAATPRT SQFTKYWTES NGVESKSLTP ALCRTPANKL KNKENVYTPK SAVKNEEYFM FPEPKTPVNK NQHKREILTT PNRYTTPSKA RNQCLKETPI KIPVNSTGTD KLMTGVISPE RRCRSVELDL NQAHMEETPK RKGAKVFGSL ERGLDKVITV LTRSKRKGSA RDGPRRLKLH YNVTTTRLVN PDQLLNEIMS ILPKKHVDFV QKGYTLKCQT QSDFGKVTMQ FELEVCQLQK PDVVGIRRQR LKGDAWVYKR LVEDILSSCK V (302-651 aa encoded by BC014039)
MELK is a serine/threonione kinase which is related to the AMPK family that has been linked to the cell cycle. MELK has also been shown to be overexpressed in cancer and has been identified has a potential target for cancer therapeutics. MELK has a broad substrate specificity; phosphorylates BCL2L14, CDC25B, MAP3K5/ASK1 and ZNF622. Acts as an activator of apoptosis by phosphorylating and activating MAP3K5/ASK1. Acts as a regulator of cell cycle, notably by mediating phosphorylation of CDC25B, promoting localization of CDC25B to the centrosome and the spindle poles during mitosis. Plays a key role in cell proliferation and carcinogenesis. Required for proliferation of embryonic and postnatal multipotent neural progenitors. Phosphorylates and inhibits BCL2L14, possibly leading to affect mammary carcinogenesis by mediating inhibition of the pro-apoptotic function of BCL2L14. Also involved in the inhibition of spliceosome assembly during mitosis by phosphorylating ZNF622, thereby contributing to its redirection to the nucleus. May also play a role in primitive hematopoiesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMELK; hPK38; Maternal embryonic leucine zipper kinase; mPK38; OTTHUMP00000046113; pEg3 kinase; Protein kinase Eg3; Protein kinase PK38; RP11-8N6.1; Tyrosine-protein kinase MELK
Gene Aliases: AI327312; HPK38; KIAA0175; MELK; mKIAA0175; MPK38; Pk38
UniProt ID: (Human) Q14680, (Mouse) Q61846
Entrez Gene ID: (Human) 9833, (Mouse) 17279
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support