Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SVVRSYCGHG IHKLFHTAPN VPHYAKNKAV GVMKSGHVFT IEPMICEGGW QDETWPDGWT AVTRDGKRSA QFEH
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
METAP1 removes the amino-terminal methionine from nascent proteins. METAP1 is required for normal progression through the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781C0419; EC 3.4.11.18; KIAA0094MAP 1; MAP 1; MAP1; metAP 1; Methionine aminopeptidase 1; Peptidase M 1
Gene Aliases: KIAA0094; MAP1A; METAP1; MetAP1A
UniProt ID: (Human) P53582
Entrez Gene ID: (Human) 23173
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support