Product References
Exercise Mitigates Endothelial Pyroptosis and Atherosclerosis by Downregulating NEAT1 Through N6-Methyladenosine Modifications.
Arteriosclerosis, thrombosis, and vascular biology
Yang Q,Chen S,Wang X,Yang X,Chen L,Huang T,Zheng Y,Zheng X,Wu X,Sun Y,Wu J
Thu Jun 01 00:00:00 EDT 2023
Role of N6-adenosine-methyltransferase subunits METTL3 and METTL14 in the biological properties of periodontal ligament cells.
Tissue & cell
Huang J,Guo C,Wang Y,Zhou Y
Thu Jun 01 00:00:00 EDT 2023
Role of N6-adenosine-methyltransferase subunits METTL3 and METTL14 in the biological properties of periodontal ligament cells.
Tissue & cell
Huang J,Guo C,Wang Y,Zhou Y
Thu Jun 01 00:00:00 EDT 2023
Role of N6-adenosine-methyltransferase subunits METTL3 and METTL14 in the biological properties of periodontal ligament cells.
Tissue & cell
Huang J,Guo C,Wang Y,Zhou Y
Thu Jun 01 00:00:00 EDT 2023
METTL14 Regulates PLAGL2/β-Catenin Signaling Axis to Promote the Development of Nonsmall Cell Lung Cancer.
Journal of oncology
Zhou Q,Lai X,Gao Y,Chen Q,Xu Y,Liu Y
Tue Mar 07 00:00:00 EST 2023
Methyltransferase-like 3 aggravates endoplasmic reticulum stress in preeclampsia by targeting TMBIM6 in YTHDF2-dependent manner.
Molecular medicine (Cambridge, Mass.)
Chen Y,Liu X,Li L,He X,Zheng F,Zhang Y,Gao H,Jin Z,Wu D,Wang Q,Tao H,Zhao Y,Liu W,Zou L
Mon Feb 06 00:00:00 EST 2023
Comprehensive analyses of molecular features, prognostic values, and regulatory functionalities of m6A-modified long non-coding RNAs in lung adenocarcinoma.
Clinical epigenetics
Ping Y,Huang J,Zhu J,Sun Z,Shang A,Chen C,Liu W,Li D
Fri Apr 07 00:00:00 EDT 2023
METTL3-mediated m6A mRNA modification was involved in cadmium-induced liver injury.
Environmental pollution (Barking, Essex : 1987)
Li W,Tan M,Wang H,Wang Z,Pang Y,Yang R,Zhong S,Pan X,Chen S,Wang Q,Li D,Xiao Y,Chen W,Chen L
Tue Aug 15 00:00:00 EDT 2023
The miR-27a-3p/FTO axis modifies hypoxia-induced malignant behaviors of glioma cells.
Acta biochimica et biophysica Sinica
Du P,Meng L,Liao X,Liu Y,Mo X,Gong M,Liao Y
Wed Jan 25 00:00:00 EST 2023
ALKBH1-mediated m1 A demethylation of METTL3 mRNA promotes the metastasis of colorectal cancer by downregulating SMAD7 expression.
Molecular oncology
Chen W,Wang H,Mi S,Shao L,Xu Z,Xue M
Wed Feb 01 00:00:00 EST 2023
Differential epitranscriptome and proteome modulation in the brain of neonatal mice exposed to isoflurane or sevoflurane.
Cell biology and toxicology
Wu Y,Yang Z,Su S,Xu X,Li Y,Li X,Gao Y,Sun D,Wan S,Pen M,Jin W,Ke C
Sun Oct 01 00:00:00 EDT 2023
m 6 A RNA methylation orchestrates transcriptional dormancy during developmental pausing.
bioRxiv : the preprint server for biology
Collignon E,Cho B,Fothergill-Robinson J,Furlan G,Ross RL,Limbach PA,Ramalho-Santos M
Wed Feb 01 00:00:00 EST 2023
Epigenetically silenced lncRNA SNAI3-AS1 promotes ferroptosis in glioma via perturbing the m6A-dependent recognition of Nrf2 mRNA mediated by SND1.
Journal of experimental & clinical cancer research : CR
Zheng J,Zhang Q,Zhao Z,Qiu Y,Zhou Y,Wu Z,Jiang C,Wang X,Jiang X
Fri May 19 00:00:00 EDT 2023
Newcastle disease virus activates methylation-related enzymes to reprogram m6A methylation in infected cells.
Veterinary microbiology
Yuan W,Hou Y,Wang Q,Lv T,Ren J,Fan L,Cai J,Xiang B,Lin Q,Liao M,Ding C,Chen L,Ren T
Thu Jun 01 00:00:00 EDT 2023
Profile of N6-methyladenosine of Pb-exposed neurons presents epitranscriptomic alterations in PI3K-AKT pathway-associated genes.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Ai S,Li D,Gu X,Xu Y,Wang Y,Wang HL,Chen XT
Tue Aug 01 00:00:00 EDT 2023
Heterogeneous Nuclear Protein U Degraded the m6A Methylated TRAF3 Transcript by YTHDF2 To Promote Porcine Epidemic Diarrhea Virus Replication.
Journal of virology
Zhou H,Yan Y,Gao J,Ma M,Liu Y,Shi X,Zhang Q,Xu X
Tue Feb 28 00:00:00 EST 2023
Macrophage membrane-coated nanovesicles for dual-targeted drug delivery to inhibit tumor and induce macrophage polarization.
Bioactive materials
Huang X,Wang L,Guo H,Zhang W
Mon May 01 00:00:00 EDT 2023
NRP1 contributes to stemness and potentiates radioresistance via WTAP-mediated m6A methylation of Bcl-2 mRNA in breast cancer.
Apoptosis : an international journal on programmed cell death
Wang Y,Zhang L,Sun XL,Lu YC,Chen S,Pei DS,Zhang LS
Wed Feb 01 00:00:00 EST 2023
Specific Regulation of m6A by SRSF7 Promotes the Progression of Glioblastoma.
Genomics, proteomics & bioinformatics
Cun Y,An S,Zheng H,Lan J,Chen W,Luo W,Yao C,Li X,Huang X,Sun X,Wu Z,Hu Y,Li Z,Zhang S,Wu G,Yang M,Tang M,Yu R,Liao X,Gao G,Zhao W,Wang J,Li J
Tue Aug 01 00:00:00 EDT 2023
METTL14-dependent maturation of pri-miR-17 regulates mitochondrial homeostasis and induces chemoresistance in colorectal cancer.
Cell death & disease
Sun K,Chen L,Li Y,Huang B,Yan Q,Wu C,Lai Q,Fang Y,Cai J,Liu Y,Chen J,Wang X,Zhu Y,Dong S,Tan J,Li A,Liu S,Zhang Y
Tue Feb 21 00:00:00 EST 2023
AS3MT facilitates NLRP3 inflammasome activation by m6A modification during arsenic-induced hepatic insulin resistance.
Cell biology and toxicology
Qiu T,Wu C,Yao X,Han Q,Wang N,Yuan W,Zhang J,Shi Y,Jiang L,Liu X,Yang G,Sun X
Sun Oct 01 00:00:00 EDT 2023
Splice site m6A methylation prevents binding of DGCR8 to suppress KRT4 pre-mRNA splicing in oral squamous cell carcinoma.
PeerJ
Li X,Fang J,Tao X,Xia J,Cheng B,Wang Y
Fri Feb 24 00:00:00 EST 2023
ZBTB7C m6A modification incurred by METTL3 aberration promotes osteosarcoma progression.
Translational research : the journal of laboratory and clinical medicine
An X,Wu W,Yang L,Dong J,Liu B,Guo J,Chen J,Guo B,Cao W,Jiang Q
Fri Sep 01 00:00:00 EDT 2023
FTO-dependent m6A modification of Plpp3 in circSCMH1-regulated vascular repair and functional recovery following stroke.
Nature communications
Li B,Xi W,Bai Y,Liu X,Zhang Y,Li L,Bian L,Liu C,Tang Y,Shen L,Yang L,Gu X,Xie J,Zhou Z,Wang Y,Yu X,Wang J,Chao J,Han B,Yao H
Mon Jan 30 00:00:00 EST 2023
Mechanism of METTL14-mediated m6A modification in non-small cell lung cancer cell resistance to cisplatin.
Journal of molecular medicine (Berlin, Germany)
Gong S,Wang S,Shao M
Thu Dec 01 00:00:00 EST 2022
Mechanism of METTL14-mediated m6A modification in non-small cell lung cancer cell resistance to cisplatin.
Journal of molecular medicine (Berlin, Germany)
Gong S,Wang S,Shao M
Thu Dec 01 00:00:00 EST 2022
Reduced N6-Methyladenosine Mediated by METTL3 Acetylation Promotes MTF1 Expression and Hepatocellular Carcinoma Cell Growth.
Chemistry & biodiversity
Yang Y,Qian Cai Q,Sheng Fu L,Wei Dong Y,Fan F,Zhong Wu X
Tue Nov 01 00:00:00 EDT 2022
Reduced N6-Methyladenosine Mediated by METTL3 Acetylation Promotes MTF1 Expression and Hepatocellular Carcinoma Cell Growth.
Chemistry & biodiversity
Yang Y,Qian Cai Q,Sheng Fu L,Wei Dong Y,Fan F,Zhong Wu X
Tue Nov 01 00:00:00 EDT 2022
Long noncoding RNA lnc-H2AFV-1 promotes cell growth by regulating aberrant m6A RNA modification in head and neck squamous cell carcinoma.
Cancer science
Chen X,Liu Y,Sun D,Sun R,Wang X,Li M,Song N,Ying J,Guo T,Jiang Y
Wed Jun 01 00:00:00 EDT 2022
YTHDF1 Negatively Regulates Treponema pallidum-Induced Inflammation in THP-1 Macrophages by Promoting SOCS3 Translation in an m6A-Dependent Manner.
Frontiers in immunology
Li Z,Teng M,Jiang Y,Zhang L,Luo X,Liao Y,Yang B
Fri Apr 22 00:00:00 EDT 2022
DDX5/METTL3-METTL14/YTHDF2 Axis Regulates Replication of Influenza A Virus.
Microbiology spectrum
Zhao L,Zhao Y,Liu Q,Huang J,Lu Y,Ping J
Wed Jun 29 00:00:00 EDT 2022
DDX5/METTL3-METTL14/YTHDF2 Axis Regulates Replication of Influenza A Virus.
Microbiology spectrum
Zhao L,Zhao Y,Liu Q,Huang J,Lu Y,Ping J
Wed Jun 29 00:00:00 EDT 2022
The expression of m6A enzymes in the hippocampus of diabetic cognitive impairment mice and the possible improvement of YTHDF1.
Brain research
Li M,Zhong X,Zhao Z,Zeng Z,Yuan Q,Xiao X,Zu X,Liu J
Tue Feb 15 00:00:00 EST 2022
METTL3 acetylation impedes cancer metastasis via fine-tuning its nuclear and cytosolic functions.
Nature communications
Li Y,He X,Lu X,Gong Z,Li Q,Zhang L,Yang R,Wu C,Huang J,Ding J,He Y,Liu W,Chen C,Cao B,Zhou D,Shi Y,Chen J,Wang C,Zhang S,Zhang J,Ye J,You H
Wed Oct 26 00:00:00 EDT 2022
METTL3 acetylation impedes cancer metastasis via fine-tuning its nuclear and cytosolic functions.
Nature communications
Li Y,He X,Lu X,Gong Z,Li Q,Zhang L,Yang R,Wu C,Huang J,Ding J,He Y,Liu W,Chen C,Cao B,Zhou D,Shi Y,Chen J,Wang C,Zhang S,Zhang J,Ye J,You H
Wed Oct 26 00:00:00 EDT 2022
METTL3 acetylation impedes cancer metastasis via fine-tuning its nuclear and cytosolic functions.
Nature communications
Li Y,He X,Lu X,Gong Z,Li Q,Zhang L,Yang R,Wu C,Huang J,Ding J,He Y,Liu W,Chen C,Cao B,Zhou D,Shi Y,Chen J,Wang C,Zhang S,Zhang J,Ye J,You H
Wed Oct 26 00:00:00 EDT 2022
Adenosine N6-methylation upregulates the expression of human CYP2B6 by altering the chromatin status.
Biochemical pharmacology
Isono M,Nakano M,Fukami T,Nakajima M
Tue Nov 01 00:00:00 EDT 2022
PDGF signaling inhibits mitophagy in glioblastoma stem cells through N6-methyladenosine.
Developmental cell
Lv D,Gimple RC,Zhong C,Wu Q,Yang K,Prager BC,Godugu B,Qiu Z,Zhao L,Zhang G,Dixit D,Lee D,Shen JZ,Li X,Xie Q,Wang X,Agnihotri S,Rich JN
Mon Jun 20 00:00:00 EDT 2022
METTL14 Regulates Intestine Cellular Senescence through m6A Modification of Lamin B Receptor.
Oxidative medicine and cellular longevity
Zhang Z,Xue M,Chen J,Wang Z,Ju F,Ni J,Sun J,Wu H,Zheng H,Lou Z,Zhang Y,Yang X,Chen S,Xi Y,Wang L
Fri Dec 30 00:00:00 EST 2022
Mettl14-Mediated m6A Modification Is Essential for Germinal Center B Cell Response.
Journal of immunology (Baltimore, Md. : 1950)
Huang H,Zhang G,Ruan GX,Li Y,Chen W,Zou J,Zhang R,Wang J,Ji SJ,Xu S,Ou X
Fri Apr 15 00:00:00 EDT 2022
Systematic analysis of the expression profile and prognostic significance of m6A regulators and PD-L1 in hepatocellular carcinoma.
Discover. Oncology
Kong F,Wang K,Wang L
Fri Nov 25 00:00:00 EST 2022
N6-methyladenosine reader YTHDF3 regulates melanoma metastasis via its 'executor'LOXL3.
Clinical and translational medicine
Shi HZ,Xiong JS,Gan L,Zhang Y,Zhang CC,Kong YQ,Miao QJ,Tian CC,Li R,Liu JQ,Zhang EJ,Bu WB,Wang Y,Cheng XF,Sun JF,Chen H
Tue Nov 01 00:00:00 EDT 2022
m6A demethylation of cytidine deaminase APOBEC3B mRNA orchestrates arsenic-induced mutagenesis.
The Journal of biological chemistry
Gao M,Qi Z,Feng W,Huang H,Xu Z,Dong Z,Xu M,Han J,Kloeber JA,Huang J,Lou Z,Liu S
Tue Feb 01 00:00:00 EST 2022
Therapeutic targeting m6A-guided miR-146a-5p signaling contributes to the melittin-induced selective suppression of bladder cancer.
Cancer letters
Yan R,Dai W,Wu R,Huang H,Shu M
Sat May 28 00:00:00 EDT 2022
The landscape of m6A regulators in esophageal cancer: molecular characteristics, immuno-oncology features, and clinical relevance.
Annals of translational medicine
Li Z,Zheng C,Huang L,Yin X,Wang Z,Liu C,Li B
Thu Dec 01 00:00:00 EST 2022
Novel prognostic biomarkers, METTL14 and YTHDF2, associated with RNA methylation in Ewing's sarcoma.
Scientific reports
Jiang J,Fan Q,Qu H,Liu C,Liang T,Chen L,Huang S,Sun X,Chen J,Chen T,Li H,Yao Y,Zhan X
Fri Apr 29 00:00:00 EDT 2022
PGE2 -EP3 axis promotes brown adipose tissue formation through stabilization of WTAP RNA methyltransferase.
The EMBO journal
Tao X,Du R,Guo S,Feng X,Yu T,OuYang Q,Chen Q,Fan X,Wang X,Guo C,Li X,Xue F,Chen S,Tong M,Lazarus M,Zuo S,Yu Y,Shen Y
Tue Aug 16 00:00:00 EDT 2022
The genomic landscape of cholangiocarcinoma reveals the disruption of post-transcriptional modifiers.
Nature communications
Zhang Y,Ma Z,Li C,Wang C,Jiang W,Chang J,Han S,Lu Z,Shao Z,Wang Y,Wang H,Jiao C,Wang D,Wu X,Shen H,Wang X,Hu Z,Li X
Wed Jun 01 00:00:00 EDT 2022
Expression pattern of alkB homolog 5 in goat testis and its role in spermatogonial stem cells.
Cell and tissue research
Liu Z,Cai Y,Deng M,Li D,Leng Q,Shi L,Tang Y,Wang F,Wan Y
Sat Jan 01 00:00:00 EST 2022
The RNA m6A reader YTHDF2 controls NK cell antitumor and antiviral immunity.
The Journal of experimental medicine
Ma S,Yan J,Barr T,Zhang J,Chen Z,Wang LS,Sun JC,Chen J,Caligiuri MA,Yu J
Mon Aug 02 00:00:00 EDT 2021
Stimulation of Let-7 Maturation by Metformin Improved the Response to Tyrosine Kinase Inhibitor Therapy in an m6A Dependent Manner.
Frontiers in oncology
Li K,Gao S,Ma L,Sun Y,Peng ZY,Wu J,Du N,Ren H,Tang SC,Sun X
Tue Jan 25 00:00:00 EST 2022
Isorhapontigenin (ISO) inhibits EMT through FOXO3A/METTL14/VIMENTIN pathway in bladder cancer cells.
Cancer letters
Zhang N,Hua X,Tu H,Li J,Zhang Z,Max C
Mon Nov 01 00:00:00 EDT 2021
The N6-methyladenosine modification posttranscriptionally regulates hepatic UGT2B7 expression.
Biochemical pharmacology
Ondo K,Isono M,Nakano M,Hashiba S,Fukami T,Nakajima M
Thu Jul 01 00:00:00 EDT 2021
ALKBH5-mediated m6A demethylation of KCNK15-AS1 inhibits pancreatic cancer progression via regulating KCNK15 and PTEN/AKT signaling.
Cell death & disease
He Y,Yue H,Cheng Y,Ding Z,Xu Z,Lv C,Wang Z,Wang J,Yin C,Hao H,Chen C
Wed Dec 01 00:00:00 EST 2021
Oncogenic AURKA-enhanced N6-methyladenosine modification increases DROSHA mRNA stability to transactivate STC1 in breast cancer stem-like cells.
Cell research
Peng F,Xu J,Cui B,Liang Q,Zeng S,He B,Zou H,Li M,Zhao H,Meng Y,Chen J,Liu B,Lv S,Chu P,An F,Wang Z,Huang J,Zhan Y,Liao Y,Lu J,Xu L,Zhang J,Sun Z,Li Z,Wang F,Lam EW,Liu Q
Mon Mar 01 00:00:00 EST 2021
Knockdown of METTL14 suppresses the malignant progression of non-small cell lung cancer by reducing Twist expression.
Oncology letters
Yang F,Yuan WQ,Li J,Luo YQ
Wed Dec 01 00:00:00 EST 2021
YTHDC1-mediated VPS25 regulates cell cycle by targeting JAK-STAT signaling in human glioma cells.
Cancer cell international
Zhu X,Yang H,Zhang M,Wu X,Jiang L,Liu X,Lv K
Sat Dec 04 00:00:00 EST 2021
MMP24 Contributes to Neuropathic Pain in an FTO-Dependent Manner in the Spinal Cord Neurons.
Frontiers in pharmacology
Ma L,Huang Y,Zhang F,Gao DS,Sun N,Ren J,Xia S,Li J,Peng X,Yu L,Jiang BC,Yan M
Tue May 18 00:00:00 EDT 2021
m6A modification impacts hepatic drug and lipid metabolism properties by regulating carboxylesterase 2.
Biochemical pharmacology
Takemoto S,Nakano M,Fukami T,Nakajima M
Mon Nov 01 00:00:00 EDT 2021
Identification and Validation of m6A-Related lncRNA Signature as Potential Predictive Biomarkers in Breast Cancer.
Frontiers in oncology
Lv W,Wang Y,Zhao C,Tan Y,Xiong M,Yi Y,He X,Ren Y,Wu Y,Zhang Q
Tue Nov 02 00:00:00 EDT 2021
Identification and Validation of m6A-Related lncRNA Signature as Potential Predictive Biomarkers in Breast Cancer.
Frontiers in oncology
Lv W,Wang Y,Zhao C,Tan Y,Xiong M,Yi Y,He X,Ren Y,Wu Y,Zhang Q
Tue Nov 02 00:00:00 EDT 2021
M6A "Writer" Gene METTL14: A Favorable Prognostic Biomarker and Correlated With Immune Infiltrates in Rectal Cancer.
Frontiers in oncology
Cai C,Long J,Huang Q,Han Y,Peng Y,Guo C,Liu S,Chen Y,Shen E,Long K,Wang X,Yu J,Shen H,Zeng S
Tue Jul 06 00:00:00 EDT 2021
Methylation and Expression of the Exercise-Related TLR1 Gene Is Associated With Low Grade Glioma Prognosis and Outcome.
Frontiers in molecular biosciences
Lu L,Hu Y,Wang C,Jiang F,Wu C
Tue Dec 07 00:00:00 EST 2021
The m6A methylome of SARS-CoV-2 in host cells.
Cell research
Liu J,Xu YP,Li K,Ye Q,Zhou HY,Sun H,Li X,Yu L,Deng YQ,Li RT,Cheng ML,He B,Zhou J,Li XF,Wu A,Yi C,Qin CF
Thu Apr 01 00:00:00 EDT 2021
AMD1 upregulates hepatocellular carcinoma cells stemness by FTO mediated mRNA demethylation.
Clinical and translational medicine
Bian X,Shi D,Xing K,Zhou H,Lu L,Yu D,Wu W
Mon Mar 01 00:00:00 EST 2021
mTORC1 stimulates cell growth through SAM synthesis and m6A mRNA-dependent control of protein synthesis.
Molecular cell
Villa E,Sahu U,O'Hara BP,Ali ES,Helmin KA,Asara JM,Gao P,Singer BD,Ben-Sahra I
Thu May 20 00:00:00 EDT 2021
Analysis of METTL3 and METTL14 in hepatocellular carcinoma.
Aging
Liu X,Qin J,Gao T,Li C,Chen X,Zeng K,Xu M,He B,Pan B,Xu X,Pan Y,Sun H,Xu T,Wang S
Fri Nov 06 00:00:00 EST 2020
Expression and Prognostic Significance of m6A-Related Genes in Lung Adenocarcinoma.
Medical science monitor : international medical journal of experimental and clinical research
Zhang Y,Liu X,Liu L,Li J,Hu Q,Sun R
Sat Feb 22 00:00:00 EST 2020
Alteration of N6-methyladenosine epitranscriptome profile in unilateral ureteral obstructive nephropathy.
Epigenomics
Li X,Fan X,Yin X,Liu H,Yang Y
Wed Jul 01 00:00:00 EDT 2020
METTL14 Inhibits Hepatocellular Carcinoma Metastasis Through Regulating EGFR/PI3K/AKT Signaling Pathway in an m6A-Dependent Manner.
Cancer management and research
Shi Y,Zhuang Y,Zhang J,Chen M,Wu S
Tue Apr 19 00:00:00 EDT 2022
METTL14 Inhibits Hepatocellular Carcinoma Metastasis Through Regulating EGFR/PI3K/AKT Signaling Pathway in an m6A-Dependent Manner.
Cancer management and research
Shi Y,Zhuang Y,Zhang J,Chen M,Wu S
Tue Apr 19 00:00:00 EDT 2022
Bone-derived mesenchymal stem cells alleviate compression-induced apoptosis of nucleus pulposus cells by N6 methyladenosine of autophagy.
Cell death & disease
Li G,Song Y,Liao Z,Wang K,Luo R,Lu S,Zhao K,Feng X,Liang H,Ma L,Wang B,Ke W,Yin H,Zhan S,Li S,Wu X,Zhang Y,Yang C
Thu Feb 06 00:00:00 EST 2020
The piRNA CHAPIR regulates cardiac hypertrophy by controlling METTL3-dependent N6-methyladenosine methylation of Parp10 mRNA.
Nature cell biology
Gao XQ,Zhang YH,Liu F,Ponnusamy M,Zhao XM,Zhou LY,Zhai M,Liu CY,Li XM,Wang M,Shan C,Shan PP,Wang Y,Dong YH,Qian LL,Yu T,Ju J,Wang T,Wang K,Chen XZ,Wang YH,Zhang J,Li PF,Wang K
Sun Nov 01 00:00:00 EDT 2020
The piRNA CHAPIR regulates cardiac hypertrophy by controlling METTL3-dependent N6-methyladenosine methylation of Parp10 mRNA.
Nature cell biology
Gao XQ,Zhang YH,Liu F,Ponnusamy M,Zhao XM,Zhou LY,Zhai M,Liu CY,Li XM,Wang M,Shan C,Shan PP,Wang Y,Dong YH,Qian LL,Yu T,Ju J,Wang T,Wang K,Chen XZ,Wang YH,Zhang J,Li PF,Wang K
Sun Nov 01 00:00:00 EDT 2020
AMPK facilitates intestinal long-chain fatty acid uptake by manipulating CD36 expression and translocation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Wu W,Wang S,Liu Q,Shan T,Wang X,Feng J,Wang Y
Wed Apr 01 00:00:00 EDT 2020
m6A mRNA Methylation Controls Functional Maturation in Neonatal Murine β-Cells.
Diabetes
Wang Y,Sun J,Lin Z,Zhang W,Wang S,Wang W,Wang Q,Ning G
Sat Aug 01 00:00:00 EDT 2020
m6A mRNA Methylation Controls Functional Maturation in Neonatal Murine β-Cells.
Diabetes
Wang Y,Sun J,Lin Z,Zhang W,Wang S,Wang W,Wang Q,Ning G
Sat Aug 01 00:00:00 EDT 2020
m6A mRNA Methylation Controls Functional Maturation in Neonatal Murine β-Cells.
Diabetes
Wang Y,Sun J,Lin Z,Zhang W,Wang S,Wang W,Wang Q,Ning G
Sat Aug 01 00:00:00 EDT 2020
m6A mRNA Methylation Controls Functional Maturation in Neonatal Murine β-Cells.
Diabetes
Wang Y,Sun J,Lin Z,Zhang W,Wang S,Wang W,Wang Q,Ning G
Sat Aug 01 00:00:00 EDT 2020
ANKLE1 N6 -Methyladenosine-related variant is associated with colorectal cancer risk by maintaining the genomic stability.
International journal of cancer
Tian J,Ying P,Ke J,Zhu Y,Yang Y,Gong Y,Zou D,Peng X,Yang N,Wang X,Mei S,Zhang Y,Wang C,Zhong R,Chang J,Miao X
Mon Jun 15 00:00:00 EDT 2020
Knockdown of METTL14 inhibits the growth and invasion of cervical cancer.
Translational cancer research
Geng F,Fan MJ,Li J,Liang SM,Li CY,Li N
Tue Oct 01 00:00:00 EDT 2019
Viral N6-methyladenosine upregulates replication and pathogenesis of human respiratory syncytial virus.
Nature communications
Xue M,Zhao BS,Zhang Z,Lu M,Harder O,Chen P,Lu Z,Li A,Ma Y,Xu Y,Liang X,Zhou J,Niewiesk S,Peeples ME,He C,Li J
Wed Oct 09 00:00:00 EDT 2019
EBV epitranscriptome reprogramming by METTL14 is critical for viral-associated tumorigenesis.
PLoS pathogens
Lang F,Singh RK,Pei Y,Zhang S,Sun K,Robertson ES
Sat Jun 01 00:00:00 EDT 2019
EBV epitranscriptome reprogramming by METTL14 is critical for viral-associated tumorigenesis.
PLoS pathogens
Lang F,Singh RK,Pei Y,Zhang S,Sun K,Robertson ES
Sat Jun 01 00:00:00 EDT 2019
Epitranscriptomic m6A Regulation of Axon Regeneration in the Adult Mammalian Nervous System.
Neuron
Weng YL,Wang X,An R,Cassin J,Vissers C,Liu Y,Liu Y,Xu T,Wang X,Wong SZH,Joseph J,Dore LC,Dong Q,Zheng W,Jin P,Wu H,Shen B,Zhuang X,He C,Liu K,Song H,Ming GL
Wed Jan 17 00:00:00 EST 2018