Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: RSWNMDSRLQE IRERQKLRRQ LLAQQLGAES ADSIGAVLNS KDEQREIAET RETCRASYDT SAPNAKRKYL DEGETDEDKM EEYKDELEMQ QDEE
Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
KIAA1627 is a probable methyltransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMETTL14; Methyltransferase-like protein 14; N6-adenosine-methyltransferase non-catalytic subunit
Gene Aliases: KIAA1627; METTL14
UniProt ID: (Human) Q9HCE5
Entrez Gene ID: (Human) 57721
Molecular Function:
RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support