Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSDTWSSIQA HKKQLDSLRE RLQRRRKQDS GHLDLRNPEA ALSPTFRSDS PVPTAPTSGG PKPSTASAVP ELATDPELEK KLLHHLSDLA LTLPTDAVSI CLAISTPDAP ATQDGVESLL QKFAAQELIE VKRGLLQDDA HPTLVTYADH SKLSAMMGAV AEKKGPGEVA GTVTGQKRRA EQDSTTVAAF ASSLVSGLNS SASEPAKEPA KKSRKHAASD VDLEIESLLN QQSTKEQQSK KVSQEILELL NTTTAKEQSI VEKFRSRGRA QVQEFCDYGT KEECMKASDA DRPCRKLHFR RIINKHTDES LGDCSFLNTC FHMDTCKYVH YEIDACMDSE APGSKDHTPS QELALTQSVG GDSSADRLFP PQWICCDIRY LDVSILGKFA VVMADPPWDI HMELPYGTLT DDEMRRLNIP VLQDDGFLFL WVTGRAMELG RECLNLWGYE RVDEIIWVKT NQLQRIIRTG RTGHWLNHGK EHCLVGVKGN PQGFNQGLDC DVIVAEVRST SHKPDEIYGM IERLSPGTRK IELFGRPHNV QPNWITLGNQ LDGIHLLDPD VVARFKQRYP DGIISKPKNL
METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; hMETTL3; Methyltransferase-like protein 3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MT-A70; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase catalytic subunit
Gene Aliases: IME4; M6A; METTL3; MT-A70; MTA70; Spo8
UniProt ID: (Human) Q86U44
Entrez Gene ID: (Human) 56339
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support