Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKALLALPL LLLLSTPPCA PQVSGIRGDA LERFCLQQPL DCDDIYAQGY QSDGVYLIYP SGPSVPVPVF CDMTTEGGKW TVFQKRFNGS VSFFRGWNDY KLGFGRADGE YWLGLQNMHL LTLKQKYELR VDLEDFENNT AYAKYADFSI SPNAVSAEED GYTLFVAGFE DGGVGDSLSY HSGQKFSTFD RDQDLFVQNC AALSSGAFWF RSCHFANLNG FYLGGSHLSY ANGINWAQWK GFYYSLKRTE MKIRRA (1-255 aa encoded by B C062415)
MFAP4 is a protein with similarity to a bovine microfibril-associated protein. The protein has binding specificities for both collagen and carbohydrate. It is thought to be an extracellular matrix protein which is involved in cell adhesion or intercellular interactions. The gene is located within the Smith-Magenis syndrome region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Microfibril-associated glycoprotein 4; microfibrillar-associated protein 4
Gene Aliases: 1110007F23Rik; Magp-36; MFAP4
UniProt ID: (Human) P55083, (Mouse) Q9D1H9
Entrez Gene ID: (Human) 4239, (Mouse) 76293, (Rat) 287382
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support