Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: SGIRGDALER FCLQQPLDCD DIYAQGYQSD GVYLIYPSGP SVPVPVFCDM TTEGGKWTVF QKRFNGSVSF FRGWNDYKLG FGRADGEYWL GLQNMHLLTL
MFAP4 is a protein with similarity to a bovine microfibril-associated protein. The protein has binding specificities for both collagen and carbohydrate. It is thought to be an extracellular matrix protein which is involved in cell adhesion or intercellular interactions. The gene is located within the Smith-Magenis syndrome region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Microfibril-associated glycoprotein 4; microfibrillar-associated protein 4
Gene Aliases: MFAP4
UniProt ID: (Human) P55083
Entrez Gene ID: (Human) 4239
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support