Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQCRLPRGLA GALLTLLCMG LLCLRYHLNL SPQRVQGTPE LSQPNPGPPK LQLHDVFIAV KTTRAFHRLR LELLLDTWVS RTREQTFVFT DSPDKGLQER LGSHLVVTNC SAEHSHPALS CKMAAEFDTF LASGLRWFCH VDDDNYVNPR ALLQLLRAFP LARDVYVGRP SLNRPIHASE PQPHNRTRLV QFWFATGGAG FCINRKLALK MAPWASGSRF MDTSALIRLP DDCTMGYIIE CKLGGRLQPS PLFHSHLETL QLLRTAQLPE QVTLSYGVFE GKLNVIKLQG PFSPEEDPSR FRSLHCLLYP DTPWCPQLGA R
This gene is a member of the fringe gene family which also includes radical and lunatic fringe genes. They all encode evolutionarily conserved secreted proteins that act in the Notch receptor pathway to demarcate boundaries during embryonic development. While their genomic structure is distinct from other glycosyltransferases, fringe proteins have a fucose-specific beta-1,3-N-acetylglucosaminyltransferase activity that leads to elongation of O-linked fucose residues on Notch, which alters Notch signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,3-N-acetylglucosaminyltransferase manic fringe; EC 2.4.1.222; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases: MFNG
UniProt ID: (Human) O00587
Entrez Gene ID: (Human) 4242
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support