Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: QQLRDRLLAN GFNECILLFS DLPEIDIERC VRESINLFCW TPKSATYRQH AQPPKPSSDS SGGRSSAPYF SAECPDPPKT DLSRESIPLN DLKSEVSPRI
Involved in the modulation of mTOR signaling and expression of mTOR complex components. Involved in the regulation of cell proliferation and growth. Involved in the control of actin-cytoskeleton organization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TBC domain-containing protein kinase-like protein; TBCK
Gene Aliases: HSPC302; IHPRF3; TBCK; TBCKL
UniProt ID: (Human) Q8TEA7
Entrez Gene ID: (Human) 93627
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support