Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: AELGKKYCVY CLAEVSPLRF RCTECQDIEL CPECFSAGAE IGHHRRYHGY QLVDGGRFTL WGPEAEGGWT SREEQLLLDA IEQFGFGNWE DMAAHVGASR TPQEVMEHY
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADA2-beta; ADA2-like protein beta; Transcriptional adapter 2-beta; transcriptional adaptor 2 (ADA2 homolog, yeast)-beta
Gene Aliases: AA474455; ADA2(beta); ADA2B; Gm145; RGD1561605; TADA2B
UniProt ID: (Human) Q86TJ2
Entrez Gene ID: (Human) 93624, (Mouse) 231151, (Rat) 289717
Molecular Function:
DNA-binding transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support