Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAARGRRAEP QGREAPGPAG GGGGGSRWAE SGSGTSPESG DEEVSGAGSS PVSGGVNLFA NDGSFLELFK RKMEEEQRQR QEEPPPGPQR PDQSAAAAGP GDPKRKGGPG STLSFVGKRR GGNKLALKTG IVAKKQKTED EVLTSKGDAW AKYMAEVKKY KAHQCGDDDK TRPLVK
C19orf43, also named as MGC2803 and fSAP18, is a protein coding gene. Exoribonuclease that is part of the telomerase RNA 3' end processing complex and which has the ability to all four unpaired RNA nucleotides from 5' end or 3' end with higher efficiency for purine bases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exoribonuclease TRIR; My029 protein; Telomerase RNA component interacting RNase
Gene Aliases: C19orf43; fSAP18; TRIR
UniProt ID: (Human) Q9BQ61
Entrez Gene ID: (Human) 79002
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support