Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTLEAIRYSR GSLQILDQLL LPKQSRYEAV GSVHQAWEAI RAMKVRGAPA IALVGCLSLA VELQAGAGGP GLAALVAFVR DKLSFLVTAR PTAVNMARAA RDLADVAARE AEREGATEEA VRERVICCTE DMLEKDLRDN RSIGDLGARH LLERVAPSGG KVTVLTHCNT GALATAGYGT ALGVIRSLHS LGRLEHAFCT ETRPYNQGAR LTAFELVYEQ IPATLITDSM VAAAMAHRGV SAVVVGADRV VANGDTANKV GTYQLAIVAK HHGIPFYVAA PSSSCDLRLE TGKEIIIEER PGQELTDVNG VRIAAPGIGV WNPAFDVTPH DLITGGIITE LGVFAPEELR TALTTTISSR DGTLDGPQM
MGC3207 catalyzes the interconversion of methylthioribose-1-phosphate (MTR-1-P) into methylthioribulose-1-phosphate (MTRu-1-P). Independently from catalytic activity, MGC3207 promotes cell invasion in response to constitutive RhoA activation by promoting FAK tyrosine phosphorylation and stress fiber turnover.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: M1Pi; Mediator of RhoA-dependent invasion; Methylthioribose-1-phosphate isomerase; methylthioribose-1-phosphate isomerase homolog; MGC3207; MRI1; MTR-1-P isomerase; S-methyl-5-thioribose-1-phosphate isomerase; S-methyl-5-thioribose-1-phosphate isomerase 1; Translation initiation factor eIF-2B subunit alpha/beta/delta-like protein
Gene Aliases: MRDI; MRI1; MTNA; UNQ6390/PRO21135; Ypr118w
UniProt ID: (Human) Q9BV20
Entrez Gene ID: (Human) 84245
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support