Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects the 35-40 kDa protein.
Immunogen sequence: MKMKLFQTI CRQLRSSKFS VESAALVAFS TSSYSCGRKK KVNPYEEVDQ EKYSNLVQSV LSSRGVAQTP GSVEEDALLC GPVSKHKLPN QGEDRRVPQN WFPIFNPERS DKPNASDPSV PLKIPLQRNV IPSVTRVLQQ TMTKQQVFLL ERWKQRMILE LGEDGFKEYT SNVFLQGKRF HEALESILSP QETLKERDEN LLKSGYIESV QHILKDVSGV RALESAVQHE TLNYIGLLDC VAEYQGKLCV IDWKTSEKPK PFIQSTFDNP LQVVAYMGAM NHDTNYSFQV QCGLIVVAYK DGSPAHPHFM DAELCSQYWT KWLLRLEEYT EKKKNQNIQK PEYSE (1-344 aa encoded by BC016869)
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGME1; Mitochondrial genome maintenance exonuclease 1
Gene Aliases: 8430406I07Rik; AI426476; bA504H3.4; C20orf72; DDK1; MGME1; MTDPS11
UniProt ID: (Human) Q9BQP7, (Mouse) Q9CXC3
Entrez Gene ID: (Human) 92667, (Mouse) 74528
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support