Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MGP is associates with the organic matrix of bone and cartilage. MGP is thought to act as an inhibitor of bone formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell growth-inhibiting gene 36 protein; MATRIX; Matrix Gla protein; MGP
Gene Aliases: GIG36; MGLAP; MGP; NTI
UniProt ID: (Human) Q6ICN5
Entrez Gene ID: (Human) 4256
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support