Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: PPPLGGAELA LRESSSPESF ITEEVDESSS PQQGTRAASI ENVLQDSSPE HCGRGPPADI YLPGRPTSME TAHGLATTSP TWPPLGGPSP DPSAAELTPL
Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase MGRN1; mahogunin ring finger 1, E3 ubiquitin protein ligase; Mahogunin RING finger protein 1; mahogunin, ring finger 1; probable E3 ubiquitin-protein ligase MGRN1; RING finger protein 156; RING-type E3 ubiquitin transferase MGRN1
Gene Aliases: KIAA0544; MGRN1; RNF156
UniProt ID: (Human) O60291
Entrez Gene ID: (Human) 23295
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support