Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: NLTVLSWDGS VQSGFLTEVH LDGQPFLRCD RQKCRAKPQG QWAEDVLGNK TWDRETRDLT GNGKDLRMTL AHIKDQKEGL HSLQEIRVCE IHEDNSTRSS QHFYYDGELF LSQNLETKEW TMPQSSRAQT LAMNVRNFLK EDAMKTKTHY HAMHADCLQE LRRYLKSGVV LRRTVPPMVN VTRSEASEGN ITVTCRASGF YPWNITLSWR QDGVSLSHDT QQWGDVLPDG NGTYQTWVAT RICQGEEQRF TCYMEHSGNH STHPVPSGKV
Major histocompatibility complex (MHC) class I proteins are ubiquitously expressed and mediate the recognition of intracellular antigens by cytotoxic T cells. A related family, termed the MHC class I chain-related (MIC) proteins are recognized by NKG2D, a receptor on NK and T cells, and promote anti-tumor activity. MICA, a member of the MIC family, is widely expressed on many tumors, and it is the MICA/NKG2D interaction that is thought to stimulate the anti-tumor reactivity by T lymphocytes. Both MICA and MICB mRNA are widely expressed in normal tissues, with MICA being present in virtually every tissue except the nervous system, suggesting that MIC protein expression may only be one component of the anti-tumor activity of the immune system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAMA-345G11.2; HLA class I antigen; MHC class I chain-related protein A; MHC class I polypeptide-related sequence A; MHC class I related sequence A; MIC-A; MICA; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence A
Gene Aliases: MIC-A; MICA; PERB11.1
UniProt ID: (Human) Q9TQA4
Entrez Gene ID: (Human) 100507436
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support