Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HRSCFRCHTC EATLWPGGYE QHPGDGHFYC LQHLPQTDHK AEGSDRGPES PELPTPSENS MPPGLSTPTA SQEGAGPVPD PSQPTRRQIR LSSPERQRLS SLNLTPDPEM EPPPKPPRSC SALARHALES SFVGWGLPVQ SPQALVAMEK EEKESPFSSE EEEEDVPLDS DVEQALQTFA KTSGTMNNYP TWRRTLLRRA KEEEMKRFCK AQTIQRRLNE IEAALRELEA EGVKLELALR RQSSSPEQQK KLWVGQLLQL VDKKNSLVAE EAELMITVQE LNLEEKQWQL DQELRGYMNR EENLKTAADR QAEDQVLRKL VDLVNQRDAL IRFQEERRLS ELALGTGAQG
May be a cytoskeletal regulator that connects NEDD9 to intermediate filaments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [F-actin]-monooxygenase MICAL1; CasL interacting molecule MICAL; MICAL-1; microtubule associated monoxygenase, calponin and LIM domain containing 1; mMical1; Molecule interacting with CasL protein 1; NEDD9 interacting protein with calponin homology and LIM domains; NEDD9-interacting protein with calponin homology and LIM domains; protein-methionine sulfoxide oxidase MICAL1
Gene Aliases: MICAL; MICAL-1; MICAL1; NICAL
UniProt ID: (Human) Q8TDZ2, (Mouse) Q8VDP3, (Rat) D3ZBP4
Entrez Gene ID: (Human) 64780, (Mouse) 171580, (Rat) 294520
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support