Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKVTVGPDP SLVYRPDVDP EVAKDKASFR NYTSGPLLDR VFTTYKLMHT HQTVDFVRSK HAQFGGFSYK KMTVMEAVDL LDGLVDESDP DVDFPNSFHA FQTAEGIRKA HPDKDWFHLV GLLHDLGKVL ALFGEPQWAV VGDTFPVGCR PQASVVFCDS TFQDNPDLQD PRYSTELGMY QPHCGLDRVL MSWGHDEYMY QVMKFNKFSL PPEAFYMIRF HSFYPWHTGR DYQQLCSQQD LAMLPWVREF NKFDLYTKCP DLPDVDKLRP YYQGLIDKYC PGILSW (1-285 aa encoded by B C073848)
MIOX (Myo-Inositol Oxygenase) is a Protein Coding gene. Among its related pathways are superpathway of inositol phosphate compounds and Porphyrin and chlorophyll metabolism. Gene Ontology (GO) annotations related to this gene include iron ion binding and oxidoreductase activity, acting on NAD(P)H.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aldehyde reductase (aldose reductase) like 6; aldehyde reductase (aldose reductase)-like 6; aldehyde reductase 6, renal; Aldehyde reductase-like 6; Inositol oxygenase; Kidney-specific protein 32; MI oxygenase; Myo-inositol oxygenase; renal-specific oxido-reducatse; renal-specific oxido-reductase; Renal-specific oxidoreductase
Gene Aliases: 0610009I10Rik; AI314022; ALDRL6; C85427; KSP32; MIOX; RSOR
UniProt ID: (Human) Q9UGB7, (Rat) Q9QXN4, (Mouse) Q9QXN5
Entrez Gene ID: (Human) 55586, (Rat) 252899, (Mouse) 56727
Molecular Function: oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support