Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TGAGMNPAR SFAPAILTGN FTNHWVYWVG PIIGGGLGSL LYDFLLFPRL KSISERLSVL KGAKPDVSNG QPEVTGEPVE LNTQAL (179-263 aa encoded by BC074913)
AQP0 is the most abundant endogenous protein in the plasma membrane of lens fiber cells where it functions not only as a water pore, but it is also involved in fiber-fiber adhesion and is crucial for fiber cell structure and organization. AQP0 contains an additional pore constriction, not seen in any other aquaporin structures, which may be responsible for pore gating. The closed AQP0 pore holds just three water molecules, which are spaced too far apart to form hydrogen bonds with each other. The C-terminal domain of AQP0 undergoes extensive post-translational modification, including many truncations, during lens aging due to the actions of m-calpain, proteases or non-enzymatic mechanisms. These truncation sites may be involved in the development of cataracts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aquaporin 0; Aquaporin-0; Aquaporin0; dominant cataract; hydropic fibers; Lens fiber major intrinsic protein; lens opacity; major intrinsic protein of eye lens fiber; MIP26; MP26
Gene Aliases: AQP0; Cat; CTRCT15; Hfi; LIM1; Lop; MIP; MIP26; MP26; Palm; shrivelled; Svl
UniProt ID: (Human) P30301, (Mouse) P51180
Entrez Gene ID: (Human) 4284, (Mouse) 17339, (Rat) 25480
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support