Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLCVGRLGG LGARAAALPP RRAGRGSLEA GIRARRVSTS WSPVGAAFNV KPQGSRLDLF GERRGLFGVP ELSAPEGFHI AQEKALRKTE LLVDRACSTP PGPQTVLIFD ELSDSLCRVA DLADFVKIAH PEPAFREAAE EACRSIGTMV EKLNTNVDLY QSLQKLLADK KLVDSLDPET RRVAELFMFD FEISGIHLDK EKRKRAVDLN VKILDLSSTF LMGTNFPNKI EKHLLPEHIR RNFTSAGDHI IIDGLHAESP DDLVREAAYK IFLYPNAGQL KCLEELLSSR DLLAKLVGYS TFSHRALQGT IAKNPETVMQ FLEKLSDKLS ERTLKDFEMI RGMKMKLNPQ NSEVMPWD (1-357 aa encoded by BC009934)
The product of this gene performs the final step in processing a specific class of nuclear-encoded proteins targeted to the mitochondrial matrix or inner membrane. This protein is primarily involved in the maturation of oxidative phosphorylation (OXPHOS)-related proteins. This gene may contribute to the functional effects of frataxin deficiency and the clinical manifestations of Friedreich ataxia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MIP; Mitochondrial intermediate peptidase
Gene Aliases: HMIP; MIP; MIPEP
UniProt ID: (Human) Q99797
Entrez Gene ID: (Human) 4285
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support