Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Immunogen sequence: MPPLKSPAAF HEQRRSLERA RTEDYLKRKI RSRPERSELV RMHILEETSA EPSLQAKQLK LKRARLADDL NEKIAQRPGP MELVEKNILP VESSLKEAII VGQVNYPKVA DSSSFDEDSS DALSPEQPAS HESQGSVPSP LEARVSEPLL SATSASPTQV VSQLPMGRDS REMLFLAEQP PLPPPPLLPP SLTNGTTIPT AKSTPTLIKQ SQPKSASEKS QRSKKAKELK PKVKKLKYHQ YIPPDQKQDR GAPPMDSSYA KILQQQQLFL QLQILNQQQQ; Positive Samples: TE-1, HeLa, Mouse lung, Rat testis; Cellular Location: Cytoplasm, Nucleus
The protein encoded by this gene interacts with the transcription factor myocardin, a key regulator of smooth muscle cell differentiation. The encoded protein is predominantly nuclear and may help transduce signals from the cytoskeleton to the nucleus. This gene is involved in a specific translocation event that creates a fusion of this gene and the RNA-binding motif protein-15 gene. This translocation has been associated with acute megakaryocytic leukemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic SAP and coiled-coil domains; Basic SAP coiled-coil transcription activator; basic, SAP and coiled-coil domain; basic, SAP, and coiled-coil domain containing protein; megakaryoblastic leukemia (translocation) 1; megakaryoblastic leukemia (translocation) 1 homolog; megakaryoblastic leukemia 1 met; Megakaryoblastic leukemia 1 protein; Megakaryoblastic leukemia 1 protein homolog; Megakaryocytic acute leukemia protein; Megakaryocytic acute leukemia protein homolog; mixed lineage leukemia; MKL/myocardin-like protein 1; MKL1-elongated derivative of yield protein; MRTF-A; Myocardin-related transcription factor A
Gene Aliases: AMKL; BSAC; FLMKL1; KIAA1438; MAL; MELODY; MKL1; MKL1met; MRTF-A; MRTFA
UniProt ID: (Human) Q969V6, (Mouse) Q8K4J6
Entrez Gene ID: (Human) 57591, (Mouse) 223701, (Rat) 315151
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support