Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 12
The Makorin ring finger protein-1 gene (MKRN1) is a highly transcribed, intron-containing source for a family of intronless mammalian genes encoding a novel class of zinc finger proteins. Phylogenetic analyses indicate that the MKRN1 gene is the ancestral founder of this gene family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase makorin-1; RING finger protein 61; RING-type E3 ubiquitin transferase makorin-1
Gene Aliases: MKRN1; RNF61
UniProt ID: (Human) Q9UHC7
Entrez Gene ID: (Human) 23608
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support