Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MASSCAVQVK LELGHRAQVR KKPTVEGFTH DWMVFVRGPE HSNIQHFVEK VVFHLHESFP RPKRVCKDPP YKVEESGYAG FILPIEVYFK NKEEPRKVRF DYDLFLHLEG HPPVNHLRCE KLTFNNPTED FRRKLLKAGG DPNRSIHTSS SSSSSSSSSS SSSSSSSSSS SSGSSSSSSS SSSSSSSSSS TSFSKPHKLM KEHKEKPSKD SREHKSAFKE PSRDHNKSSK ESSKKPKENK PLKEEKIVPK MAFKEPKPMS KEPKPDSNLL TITSGQDKKA PSKRPPISDS EELSAKKRKK SSSEALFKSF SSAPPLILTC SADKKQIKDK SHVKMGKVKI ESETSEKKKS TLPPFDDIVD PNDSDVEENI SSKSDSEQPS PASSSSSSSS SFTPSQTRQQ GPLRSIMKDL HSDDNEEESD EVEDNDNDSE MERPVNRGGS RSRRVSLSDG SDSESSSASS PLHHEPPPPL LKTNNNQILE VKSPIKQSKS DKQIKNGECD KAYLDELVEL HRRLMTLRER HILQQIVNLI EETGHFHITN TTFDFDLCSL DKTTVRKLQS YLETSGTS
The human AF9 gene is one of the most common fusion partner genes with the ALL1 gene at 11q23 (also called MLL), resulting in the t(9;11)(p22;q23). The AF9 gene is more than 100 kb, and 2 patient breakpoint cluster regions (BCRs) have been identified; BCR1 is within intron 4, previously called site A, whereas BCR2 or site B spans introns 7 and 8. Several different structural elements have been identified in AF9, including a colocalizing in vivo DNA topo II cleavage site and an in vitro DNase I hypersensitive (DNase 1 HS) site in intron 7 in BCR2. Reversibility experiments demonstrated a religation of the topo II cleavage sites. In addition, 2 scaffold associated regions (SARs) are located centromeric to the topo II and DNase I HS cleavage sites and border breakpoint regions in 2 leukemic cells lines: SAR1 is located in intron 4, whereas SAR2 encompasses parts of exons 5-7. The patient breakpoint regions of AF9 share the same structural elements as the MLL BCR. A DNA breakage and repair model for nonhomologous recombination between MLL and its partner genes, particularly AF9, has been proposed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AF-9 protein; ALL1-fused gene from chromosome 9 protein; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 3; Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein; myeloid/lymphoid or mixed-lineage leukemia; translocated to 3; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 3; Protein AF-9; YEATS domain-containing protein 3
Gene Aliases: AF9; MLLT3; YEATS3
UniProt ID: (Human) P42568
Entrez Gene ID: (Human) 4300
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support