Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Mmgt1 encodes a protein that that mediates Mg(2+) transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ER membrane protein complex subunit 5; Membrane magnesium transporter 1; membrane magnesium transporter 1 {ECO:0000250|UniProtKB:Q8K273}; mmgt1 {ECO:0000250|UniProtKB:Q8K273}; Transmembrane protein 32; transmembrane protein 32 {ECO:0000312|EMBL:AAI68927.1}
Gene Aliases: 9630048L06Rik; BC032271; EMC5; mir-5116; Mir5116; MMGT1; RGD1566339; TMEM32
UniProt ID: (Human) Q8N4V1, (Mouse) Q8K273, (Rat) B5DF51
Entrez Gene ID: (Human) 93380, (Mouse) 236792, (Rat) 302864
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support