Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the protein encoded by this gene is a member of the membrane-type MMP (MT-MMP) subfamily; each member of this subfamily contains a potential transmembrane domain suggesting that these proteins are expressed at the cell surface rather than secreted. This protein activates MMP2 protein, and this activity may be involved in tumor invasion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 3.4.24.80; matrix metallo protease; matrix metallopeptidase 14 (membrane-inserted); Matrix metalloproteinase-14; membrane type 1 metalloprotease; Membrane-type matrix metalloproteinase 1; Membrane-type-1 matrix metalloproteinase; MMP; MMP-14; MMP-X MT-MMP; MMP-X1; MMPs; MT-MMP 1; MT-MMP MT1-MMP; MT1-MMP
Gene Aliases: MMP-14; MMP-X1; MMP14; MT-MMP; MT-MMP 1; MT1-MMP; MT1MMP; MTMMP1; WNCHRS
UniProt ID: (Human) P50281
Entrez Gene ID: (Human) 4323
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support