Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: TRYGDPIQIL TGWPGIPTHN IDAFVHIWTW KRDERYFFQG NQYWRYDSDK DQALTEDEQG KSYPKLISEG FPGIPSPLDT AFYDRRQKLI YFFKESLVFA FDVNRNRV
This gene encodes a member of the matrix metalloproteinase family. Proteins in this family are involved in the breakdown of extracellular matrix for both normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, and disease processes, such as asthma and metastasis. The encoded protein may play an important role in embryogenesis, particularly in neuronal cells, as well as in lymphocyte development and survival.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: matrix metallo protease; matrix metalloproteinase 21; Matrix metalloproteinase-21; MMP-21; MMPs
Gene Aliases: HTX7; MMP-21; MMP21
UniProt ID: (Human) Q8N119
Entrez Gene ID: (Human) 118856
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support