Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen Sequence: MSFLFSSRSS KTFKPKKNIP EGSHQYELLK HAEATLGSGN LRQAVMLPEG EDLNEWIAVN TVDFFNQINM LYGTITEFCT EASCPVMSAG PRYEYHWADG
The protein encoded by this gene is a component of the Hippo signaling pathway, which controls organ size and tumor growth by enhancing apoptosis. Loss of the encoded protein results in cell proliferation and cancer formation. The encoded protein is also involved in the control of microtubule stability during cytokinesis. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MOB kinase activator 1A; MOB kinase activator 1B; Mob1 alpha; Mob1 homolog 1A; Mob1 homolog 1B; MOB1 Mps One Binder homolog A; MOB1 Mps One Binder homolog B; MOB1, Mps One Binder kinase activator-like 1A; MOB1, Mps One Binder kinase activator-like 1B; Mob1A; Mob1B; MOBKL1B/MOB4A; Mps one binder kinase activator-like 1A; Mps one binder kinase activator-like 1B; Protein Mob4B
Gene Aliases: 1110003E08Rik; 4022402H07Rik; AU015450; B230364F10; C2orf6; MATS1; MATS2; MOB1; MOB1A; MOB1B; MOB4A; MOB4B; MOBK1B; MOBKL1A; MOBKL1B
UniProt ID: (Human) Q9H8S9, (Human) Q7L9L4, (Mouse) Q921Y0, (Rat) Q3T1J9, (Mouse) Q8BPB0
Entrez Gene ID: (Human) 55233, (Human) 92597, (Mouse) 232157, (Rat) 297387, (Rat) 360920, (Mouse) 68473
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support